Learn More
Innovative Research Inc HUM/MOUSE FASL SOLUBLE RECOMB
Supplier:  Innovative Research Inc IHUMSFASLSRFCLY10UG
Encompass_Preferred
Human / Mouse FasL Soluble Recombinant Protein Fc Tag Lyophilized from Innovative Research has been recombinantly produced in HEK293 cells. This is a Lyophilized protein buffered in Contains PBS. Reconstitute with 100 and#956;l sterile water. with a purity of >95%
Catalog No. NC2588890
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.