Learn More
Vector Laboratories Human MUC5B (Sandwich ELISA) ELISA Kit, 1 Plate
SDP
Supplier:  Vector Laboratories LSF226091
Encompass
S-F22609 is a 96-well enzyme-linked immunosorbent assay (ELISA) for the Quantitative detection of Human MUC5B in samples of Plasma and Serum. It is based upon a Sandwich assay principle and can be used to detect levels of MUC5B as low as 0.38 nanograms per millilter.
Catalog No. NC2880676
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.