Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human 11S REG gamma (aa 1-14) Synthetic Peptide

Catalog No. PEP091
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP091 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP091 Supplier Invitrogen™ Supplier No. PEP091
Add to Cart
Add to Cart

Synthetic Peptide

Human 11S REG gamma (residues 1-14) Synthetic Peptide, PEP-091, from Invitrogen.

SFRS10 contains 1 RRM (RNA recognition motif) domain and belongs to the splicing factor SR family. It is a sequence-specific RNA-binding protein which participates in the control of pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 10197
Species Human
Purification Method purified
Accession Number P61289
Common Name Human 11 S REG gamma (aa 1-14)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol PSME3
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence M1 - A - S - L - L - K - V - D - Q - E - V - K - L - K(14) - C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.