Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human 14-3-3 beta/alpha Recombinant Protein
SDP

Catalog No. NB5965710UG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB5965710UG 10 μg
NB5965750UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB5965710UG Supplier Novus Biologicals™ Supplier No. NBP25965710UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-246 of Human 14-3-3 beta/alpha The Recombinant Human 14-3-3 beta/alpha Protein is derived from E. coli. The Recombinant Human 14-3-3 beta/alpha Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) PAGE
Formulation In PBS (pH7.4) containing 10% glycerol
Molecular Weight (g/mol) 30.6 kDa
Name YWHAB Recombinant Protein
Purification Method >90% by SDS-PAGE
Quantity 10 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias 14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha
Gene Symbol YWHAB
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.