Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human 14-3-3 epsilon Recombinant Protein
SDP

Catalog No. NB59654100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB59654100U 100 μg
NB5965420UG 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB59654100U Supplier Novus Biologicals™ Supplier No. NBP259654100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-255 of Human 14-3-3 epsilon The Recombinant Human 14-3-3 epsilon Protein is derived from E. coli. The Recombinant Human 14-3-3 epsilon Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) PAGE
Formulation In PBS (pH7.4) containing 1mM DTT, 10% glycerol
Molecular Weight (g/mol) 31.3 kDa
Name YWHAE Recombinant Protein
Purification Method >90% by SDS-PAGE
Quantity 100 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias 14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha
Gene Symbol YWHAE
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.