Learn More
Biomatik Corporation Human 17-Beta-Hydroxysteroid Dehydrogenase Type 14 (HSD17b14) ELISA Kit
Supplier:  Biomatik Corporation EKU02009
Encompass_Preferred
Human 17-Beta-Hydroxysteroid Dehydrogenase Type 14 (HSD17b14) ELISA Kit for detecting HSD17b14 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BPX1. Gene ID: 51171. Target Synonyms: DHRS10; RetSDR3, SDR47C1; Retinal Short-Chain Dehydrogenase/Reductase 3; Short Chain Dehydrogenase/Reductase Family 47C,Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9158
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.