Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human 26S Proteasome ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP266362 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266362 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266362 Supplier Novus Biologicals™ Supplier No. NBP266362
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human 26S Proteasome ELISA Kit (Chemiluminescence) measures human 26S Proteasome in serum, plasma and other biological fluids.

Specifications

Assay Range 62.50-4000 pg/mL
Assay Sensitivity 37.50 pg/mL
Cross Reactivity This kit recognizes Human 26S-PSM in samples. No significant cross-reactivity or interference between Human 265-PSM and analogues was observed.
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Host Species Human
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target 26S Proteasome
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.