Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 5-HT4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6545
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6545 100 μg
MAB6545SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6545 Supplier R&D Systems Supplier No. MAB6545
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

5-HT4 Monoclonal specifically detects 5-HT4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen 5-HT4
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 631769
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias 5HT4, 5-HT-4, 5-HT4R, 5-hydroxytryptamine (serotonin) receptor 4,5-hydroxytryptamine receptor 4,5-HT45-hydroxytryptamine4 receptor, cardiac 5-HT4 receptor, HTR4, Serotonin receptor 4
Gene Symbols HTR4
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human 5-HT4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3360
Test Specificity Detects human 5-HT4 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.