Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human 5'-Nucleotidase/CD73 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP266370 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266370 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266370 Supplier Novus Biologicals™ Supplier No. NBP266370

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human 5'-Nucleotidase/CD73 ELISA Kit (Chemiluminescence) measures human 5'-Nucleotidase/CD73 in serum, plasma and other biological fluids.

Specifications

Assay Range 62.50-4000 pg/mL
Assay Sensitivity 37.50 pg/mL
Cross Reactivity This kit recognizes Human 5-NT in samples. No significant cross-reactivity or interference between Human 5-NT and analogues was observed.
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 4907
Gene Symbol NT5E
Host Species Human
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target 5'-Nucleotidase/CD73
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.