Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ABCD1 Full-length ORF (NP_000024.2, 1 a.a. - 745 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89932147 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-147 25 μg
89-932-146 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-932-147 Supplier Abnova™ Supplier No. H00000215P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. This peroxisomal membrane protein is likely involved in the peroxisomal transport or catabolism of very long chain fatty acids. Defects in this gene have been identified as the underlying cause of adrenoleukodystrophy, an X-chromosome recessively inherited demyelinating disorder of the nervous system. [provided by RefSeq]

Sequence: MPVLSRPRPWRGNTLKRTAVLLALAAYGAHKVYPLVRQCLAPARGLQAPAGEPTQEASGVAAAKAGMNRVFLQRLLWLLRLLFPRVLCRETGLLALHSAALVSRTFLSVYVARLDGRLARCIVRKDPRAFGWQLLQWLLIALPATFVNSAIRYLEGQLALSFRSRLVAHAYRLYFSQQTYYRVSNMDGRLRNPDQSLTEDVVAFAASVAHLYSNLTKPLLDVAVTSYTLLRAARSRGAGTAWPSAIAGLVVFLTANVLRAFSPKFGELVAEEARRKGELRYMHSRVVANSEEIAFYGGHEVELALLQRSYQDLASQINLILLERLWYVMLEQFLMKYVWSASGLLMVAVPIITATGYSESDAEAVKKAALEKKEEELVSERTEAFTIARNLLTAAADAIERIMSSYKEVTELAGYTARVHEMFQVFEDVQRCHFKRPRELEDAQAGSGTIGRSGVRVEGPLKIRGQVVDVEQGIICENIPIVTPSGEVVVASLNIRVEEGMHLLITGPNGCGKSSLFRILGGLWPTYGGVLYKPPPQRMFYIPQRPYMSVGSLRDQVIYPDSVEDMQRKGYSEQDLEAILDVVHLHHILQREGGWEAMCDWKDVLSGGEKQRIGMARMFYHRPKYALLDECTSAVSIDVEGKIFQAAKDAGIALLSITHRPSLWKYHTHLLQFDGEGGWKFEKLDSAARLSLTEEKQRLEQQLAGIPKMQRRLQELCQILGEAVAPAHVPAPSPQGPGGLQGAST

Specifications

Accession Number NP_000024.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 215
Molecular Weight (g/mol) 109.3kDa
Name ABCD1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ABC42/ALD/ALDP/AMN
Common Name ABCD1
Gene Symbol ABCD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.