Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ABI1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10249100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10249100 100 μg
MAB10249SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10249100 Supplier R&D Systems Supplier No. MAB10249100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ABI1 Monoclonal specifically detects ABI1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen ABI1
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1013203
Conjugate Unconjugated
Dilution Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. Q8IZP0
Gene Alias Abelson interactor 1, ABI-1, abl interactor 1, Abl-binding protein 4, AblBP4, abl-interactor 1, Abl-interactor protein 1 long, E3B1, eps8 binding protein, Eps8 SH3 domain-binding protein, Eps8-binding protein, interactor protein AblBP4, nap1 binding protein, Nap1-binding protein, NAP1BP, spectrin SH3 domain binding protein 1, Spectrin SH3 domain-binding protein 1, SSH3BP, SSH3BP1
Host Species Mouse
Immunogen E. coli-derived recombinant human ABI1, Val429-Asp508, Accession # Q8IZP0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10006
Test Specificity Detects human ABI1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.