Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ABLIM1 Full-length ORF (AAH02448.1, 1 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89932231 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-231 10 ug
89-932-232 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-932-231 Supplier Abnova™ Supplier No. H00003983P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a cytoskeletal LIM protein that binds to actin filaments via a domain that is homologous to erythrocyte dematin. LIM domains, found in over 60 proteins, play key roles in the regulation of developmental pathways. LIM domains also function as protein-binding interfaces, mediating specific protein-protein interactions. The protein encoded by this gene could mediate such interactions between actin filaments and cytoplasmic targets. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MFTEGEEMYLQGSTVWHPDCKQSTKTEEKLRAKVDNEILDYKDLAAIPKVKAIYDIERPDLITYEPFYTSGYDDKQERQSLGESPRTLSPTPSAEGYQDVRDRMIHRSTSQGSINSPVYSRHSYTPTTSRSPQHFHRPDQGINIYRKPPIYKQHAALAAQSKSSEDIIKFSKFPAAQAPDPSETPKIETDHWPGPPSFAVVGPDMKRRSSGREEDDEELLRRRQLQEEQLMKLNSGLGQLILKEEMEKESRERSSLLASRYDSPINSASHIPSSKTASLPGYGRNGLHRPVSTDFAQYNSYGDVSGGVRDYQTLPDGHMPAMRMDRGVSMPNMLEPKIFPYEMLMVTNRGRNKILREVDRTRLERHLAPEVFREIFGMSIQEFDRLPLWRRNDMKKKAKLF

Specifications

Accession Number AAH02448.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3983
Molecular Weight (g/mol) 72.5kDa
Name ABLIM1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ABLIM/DKFZp781D0148/FLJ14564/KIAA0059/LIMAB1/LIMATIN/MGC1224
Common Name ABLIM1
Gene Symbol ABLIM1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.