Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ABO (NP_065202, 54 a.a. - 354 a.a. ) Full-length Recombinant Protein with His tag

Catalog No. 89932236 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-236 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-932-236 Supplier Abnova™ Supplier No. P3941
Add to Cart
Add to Cart

Used for SDS-PAGE

This gene encodes proteins related to the first discovered blood group system, ABO. Which allele is present in an individual determines the blood group. The 'O' blood group is caused by a deletion of guanine-258 near the N-terminus of the protein which results in a frameshift and translation of an almost entirely different protein. Individuals with the A, B, and AB alleles express glycosyltransferase activities that convert the H antigen into the A or B antigen. Other minor alleles have been found for this gene. [provided by RefSeq]

Sequence: MGSSHHHHHHSSGLVPRGSHMAVREPDHLQRVSLPRMVYPQPKVLTPCRKDVLVVTPWLAPIVWEGTFNIDILNEQFRLQNTTIGLTVFAIKKYVAFLKLFLETAEKHFMVGHRVHYYVFTDQPAAVPRVTLGTGRQLSVLEVRAYKRWQDVSMRRMEMISDFCERRFLSEVDYLVCVDVDMEFRDHVGVEILTPLFGTLHPGFYGSSREAFTYERRPQSQAYIPKDEGDFYYLGGFFGGSVQEVQRLTRACHQAMMVDQANGIEAVWHDESHLNKYLLRHKPTKVLSPEYLWDQQLLGWPAVLRKLRFTAVPKNHQAVRNP

Specifications

Accession Number NP_065202
Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation In 20mM Tris-HCl, 200mM NaCl, pH 8.0 (2mM dithiothreitol, 20% glycerol)
Gene ID (Entrez) 28
Molecular Weight (g/mol) 37.4kDa
Name ABO (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing Loading 3 ug protein in 15% SDS PAGE.
Quantity 50 μg
Storage Requirements Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias A3GALNT/A3GALT1/GTB/NAGAT
Common Name ABO
Gene Symbol ABO
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >85% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.