Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ACAD10 Full-length ORF (AAH15056.1, 1 a.a. - 237 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89932270 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-270 25 μg
89-932-269 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-932-270 Supplier Abnova™ Supplier No. H00080724P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the acyl-CoA dehydrogenase family of enzymes (ACADs), which participate in the beta-oxidation of fatty acids in mitochondria. The encoded enzyme contains a hydrolase domain at the N-terminal portion, a serine/threonine protein kinase catlytic domain in the central region, and a conserved ACAD domain at the C-terminus. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: MGGVLIPSPGRVAAEWEVQNRIPSGTILKALMEGGENGPWMRFMRAEITAEGFLREFGRLCSEMLKTSVPVDSFFSLLTSERVAKQFPVMTEAITQIRAKGLQTAVLSNNFYLPNQKSFLPLDRKQFDVIVESCMEGICKPDPRIYKLCLEQLGLQPSESIFLDDLGTNLKEAARLGIHTIKVNDPETAVKELEALLGFTLRVGVPNTRPVKKTMEIPKDSLQKYLKDLLGIQTTGL

Specifications

Accession Number AAH15056.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80724
Molecular Weight (g/mol) 52.8kDa
Name ACAD10 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC5601
Common Name ACAD10
Gene Symbol ACAD10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.