Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ACCN2 Full-length ORF (NP_001086.2, 1 a.a. - 528 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89932314 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-314 10 μg
89-932-315 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-932-314 Supplier Abnova™ Supplier No. H00000041P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, 2 hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. The member encoded by this gene is expressed in most if not all brain neurons, and it may be an ion channel subunit; however, its function as an ion channel remains unknown. Alternative splicing of this gene generates 2 transcript products. [provided by RefSeq]

Sequence: MELKAEEEEVGGVQPVSIQAFASSSTLHGLAHIFSYERLSLKRALWALCFLGSLAVLLCVCTERVQYYFHYHHVTKLDEVAASQLTFPAVTLCNLNEFRFSQVSKNDLYHAGELLALLNNRYEIPDTQMADEKQLEILQDKANFRSFKPKPFNMREFYDRAGHDIRDMLLSCHFRGEVCSAEDFKVVFTRYGKCYTFNSGRDGRPRLKTMKGGTGNGLEIMLDIQQDEYLPVWGETDETSFEAGIKVQIHSQDEPPFIDQLGFGVAPGFQTFVACQEQRLIYLPPPWGTCKAVTMDSDLDFFDSYSITACRIDCETRYLVENCNCRMVHMPGDAPYCTPEQYKECADPALDFLVEKDQEYCVCEMPCNLTRYGKELSMVKIPSKASAKYLAKKFNKSEQYIGENILVLDIFFEVLNYETIEQKKAYEIAGLLGDIGGQMGLFIGASILTVLELFDYAYEVIKHKLCRRGKCQKEAKRSSADKGVALSLDDVKRHNPCESLRGHPAGMTYAANILPHHPARGTFEDFTC

Specifications

Accession Number NP_001086.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 41
Molecular Weight (g/mol) 86.3kDa
Name ACCN2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ASIC/ASIC1/ASIC1A/BNaC2/hBNaC2
Common Name ACCN2
Gene Symbol ACCN2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.