Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ACCN5 Full-length ORF (AAI46498.1, 1 a.a. - 505 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89932316 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-316 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-932-316 Supplier Abnova™ Supplier No. H00051802P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene belongs to the amiloride-sensitive Na+ channel and degenerin (NaC/DEG) family, members of which have been identified in many animal species ranging from the nematode to human. The amiloride-sensitive Na(+) channel encoded by this gene is primarily expressed in the small intestine, however, its exact function is not known. [provided by RefSeq]

Sequence: MEQTEKSKVYAENGLLEKIKLCLSKKPLPSPTERKKFDHDFAISTSFHGIHNIVQNRSKIRRVLWLVVVLGSVSLVTWQIYIRLLNYFTWPTTTSIEVQYVEKMEFPAVTFCNLNRFQTDAVAKFGVIFFLWHIVSKVLHLQEITANSTGSREATDFAASHQNFSIVEFIRNKGFYLNNSTLLDCEFFGKPCSPKDFAHVFTEYGNCFTFNHGETLQAKRKVSVSGRGLSLLFNVNQEAFTDNPALGFVDAGIIFVIHSPKKVPQFDGLGLLSPVGMHARVTIRQVKTVHQEYPWGECNPNIKLQNFSSYSTSGCLKECKAQHIKKQCGCVPFLLPGYGIECDLQKYFSCVSPVLDHIEFKDLCTVGTHNSSCPVSCEEIEYPATISYSSFPSQKALKYLSKKLNQSRKYIRENLVKIEINYSDLNYKITQQQKAVSVSELLADLGGQLGLFCGASLITIIEIIEYLFTNFYWICIFFLLKISEMTQWTPPPQNHLGNKNRIEEC

Specifications

Accession Number AAI46498.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51802
Molecular Weight (g/mol) 82.5kDa
Name ACCN5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HINAC/INAC
Common Name ACCN5
Gene Symbol ACCN5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.