Learn More
ACROBiosystems ACROBiosystems Human ACE2 / ACEH Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems AC2H52H410MG
Encompass_Preferred
Recombinant Protein;10MG;Human ACE2, His Tag (AC2-H52H4) is expressed from human 293 cells (HEK293).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 85.5 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, His Tag (Cat. No. AC2-H52H4) at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 S protein RBD, Fc Tag (Cat. No. SPD-C5255) with a linear range of 0.1-0.8 ng/mL (QC tested).;Immobilized Human ACE2, His Tag (Cat. No. AC2-H52H4) at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 S1 protein, Fc Tag (Cat. No. S1N-C5255) with a linear range of 0.4-6 ng/mL (Routinely tested).
Catalog No. 50-210-7425
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.