Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Acetylcholinesterase/ACHE DuoSet ELISA

Catalog No. DY757405
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY757405 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY757405 Supplier R&D Systems™ Supplier No. DY757405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Acetylcholinesterase/ACHE DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Acetylcholinesterase/ACHE DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Acetylcholinesterase/ACHE.

Specifications

Assay Range 250.00 to 16000 pg/mL
Product Type Solid Phase Sandwich ELISA
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 43
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Target Acetylcholinesterase/ACHE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.