Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ACOT8 Partial ORF (NP_005460.2, 108 a.a. - 215 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89958651 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-651 25 μg
89-958-650 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-958-651 Supplier Abnova™ Supplier No. H00010005Q01L

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a peroxisomal thioesterase that appears to be involved more in the oxidation of fatty acids rather than in their formation. The encoded protein can bind to the human immunodeficiency virus-1 protein Nef, and mediate Nef-induced down-regulation of CD4 in T-cells. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq]

Sequence: SVRSVKAVQHGKPIFICQASFQQAQPSPMQHQFSMPTVPPPEELLDCETLIDQYLRDPNLQKRYPLALNRIAAQEVPIEIKPVNPSPLSQLQRMEPKQMFWVRARGYI

Specifications

Accession Number NP_005460.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10005
Molecular Weight (g/mol) 37.62kDa
Name ACOT8 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen SVRSVKAVQHGKPIFICQASFQQAQPSPMQHQFSMPTVPPPEELLDCETLIDQYLRDPNLQKRYPLALNRIAAQEVPIEIKPVNPSPLSQLQRMEPKQMFWVRARGYI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HNAACTE/PTE-2/PTE1/PTE2/hACTE-III/hTE
Common Name ACOT8
Gene Symbol ACOT8
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.