Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Actin Gamma 1 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP266399 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266399 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266399 Supplier Novus Biologicals™ Supplier No. NBP266399

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human Actin Gamma 1 ELISA Kit (Chemiluminescence) measures human Actin Gamma 1 in serum, plasma and other biological fluids.

Specifications

Assay Range 15.63-1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Cross Reactivity This kit recognizes Human ACTG1 in samples. No significant cross-reactivity or interference between Human ACTG1 and analogues was observed.
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 71
Gene Symbol ACTG1
Host Species Human
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Actin Gamma 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.