Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Active Caspase-3 Quantikine ELISA Kit
Human Active Caspase-3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Supplier: R&D Systems™ KM300
This item is not returnable.
View return policy
Description
The Human Active Caspase-3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Caspase-3 in cell extracts (100 ul).Specifications
0.31 - 20ng/mL | |
HRP | |
Biotin-ZVKD-fmk modified natural and biotin-D-fmk modified recombinant active Caspase-3. This assay also detects mouse active Caspase-3. | |
836 | |
Cell Extracts (100uL) | |
Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama |
0.1ng/mL | |
Cell Extracts | |
ELISA | |
1 x 96-well Plate | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction