Learn More
Abcam Human Activin A ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1195681X96TESTS
Encompass_Preferred
Human Activin A ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Activin A in Human in Tissue Extracts, Cell culture supernatant, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2586933
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.