Learn More
Invitrogen™ Human Activin A Recombinant Protein
Description
RP-75635 contains a purified 116 amino acid fragment (aa311-426) corresponding to the mature Inhibin beta A chain protein with an N-terminal hexahistidine tag. RP-75635 may be used as a positive control in Western blot assays.
Specifications
Specifications
| Accession Number | P08476 |
| Concentration | 0.05 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | Laemmli with no preservative; pH 6.8 |
| Gene ID (Entrez) | 3624 |
| Molecular Weight (g/mol) | 17.47 kDa |
| Name | Human Activin A |
| pH Range | 6.8 |
| Quantity | 10 μg |
| Source | E. coli |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.