Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADAM22 Full-length ORF (AAH36029.1, 1 a.a. - 342 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89932556 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-556 10 μg
89-932-557 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-932-556 Supplier Abnova™ Supplier No. H00053616P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is highly expressed in the brain and may function as an integrin ligand in the brain. Alternative splicing results in several transcript variants. [provided by RefSeq]

Sequence: MQAAVAVSVPFLLLCVLGTCPPARCGQAGDASLMELEKRKENRFVERQSIVPLRLIYRSGGEDESRHDALDTRVRGDVGGPQLTHVDQASFQVDAFGTSFILDVVLNHDLLSSEYIERHIEHGGKTVEVKGGEHCYYQGHIRGNPDSFVALSTCHGLHGMFYDGNHTYLIEPEENDTTQEDFHFHSVYKSRLFEFSLDDLPSEFQQVNITPSKFILKPRPKRSKRQLRRYPRNVEEETKYIELMIVNDHLMFKKHRLSVVHTNTYAKSVVNMADLIYKDQLKTRIVLVAMETWATDNKFAISENPLITLREFMKYRRDFIKEKSDAVHLFSYVTSVMMYYFF

Specifications

Accession Number AAH36029.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 53616
Molecular Weight (g/mol) 65.9kDa
Name ADAM22 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MDC2/MGC149832
Common Name ADAM22
Gene Symbol ADAM22
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.