Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADAM7 Full-length ORF (AAI48736.1, 1 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89932568 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-568 10 μg
89-932-569 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-932-568 Supplier Abnova™ Supplier No. H00008756P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The ADAM family is composed of zinc-binding proteins that can function as adhesion proteins and/or endopeptidases. They are involved in a number of biologic processes, including fertilization, neurogenesis, muscle development, and immune response.[supplied by OMIM]

Sequence: MLPGCIFLMILLIPQVKEKFILGVEGQQLVRPKKLPLIQKRDTGHTHDDDILKTYEEELLYEIKLNRKTLVLHLLRSREFLGSNYSETFYSMKGEAFTRHPQIMDHCFYQGSIVHEYDSAASISTCNGLRGFFRINDQRYLIEPVKYSDEGEHLVFKYNLRVPYGANYSCTELNFTRKTVPGDNESEEDSKIKGIHDEKYVELFIVADDTVYRRNGHPHNKLRNRIWGMVNFVNMIYKTLNIHVTLVGIEIWTHEDKIELYSNIETTLLRFSFWQEKILKTRKDFDHVVLLSGKWLYSHVQGISYPGGMCLPYYSTSIIKDLLPDTNIIANRMAHQLGHNLGMQHDEFPCTCPSGKCVMDSDGSIPALKFSKCSQNQYHQYLKDYKPTCMLNIPFPYNFHDFQFCGNKKLDEGEECDCGPAQECTNPCCDAHTCVLKPGFTCAEGECCESCQIKKAGSICRPAKDECDFPEMCTGHSPACPKDQFRVNGFPCKNSEGYCFMGKCPTREDQCSELFDDEAIESHDICYKMNTKGNKFGYCKNKENRFLPCEEKDVRCGKIYCTGGELSSLLGEDKTYHLKDPQKNATVKCKTIFLYHDSTDIGLVASGTKCGEGMVCNNGECLNMEKVYISTNCPSQCNENPVDGHGLQCHCEEGQAPVACEETLHVTNITILVVVLVLVIVGIGVLILLVRYRKCIKLKQVQSPPTETLGVENKGYFGDEQQIRTEPILPEIHFLNKPASKDSRGIADPNQSAK

Specifications

Accession Number AAI48736.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8756
Molecular Weight (g/mol) 109.89kDa
Name ADAM7 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias EAPI/GP-83
Common Name ADAM7
Gene Symbol ADAM7
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.