Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ADAMTS-13 ELISA Kit

Catalog No. EH12RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH12RB 96 Tests
EH12RBX5 5 x 96 Tests
EH12RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH12RB Supplier Invitrogen™ Supplier No. EH12RB
Add to Cart
Add to Cart

ELISA

Human ADAMTS-13 quantitates human ADAMTS-13 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ADAMTS-13.

ADAMTS13 is a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme is the von Willebrand Factor (vWF)-cleaving protease, which is responsible for cleaving at the site of Tyr842-Met843 of the vWF molecule. A deficiency of this enzyme is associated with thrombotic thrombocytopenic purpura.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 13; A disintegrin and metalloproteinase with thrombospondin motifs 13-like; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 13; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 13; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 13; ADAM metallopeptidase with thrombospondin type 1 motif, 13; ADAMs; ADAM-TS 13; Adamts13; ADAM-TS13; ADAMTS-13; C9orf8; Gm710; LOC102554393; metalloendopeptidases; UNQ6102/PRO20085; von Willebrand factor-cleaving protease; vWF-cleaving protease; VWFCP; vWF-CP; vWF-CP mRNA for von Willebrand factor-cleaving
Assay Range 7.5 to 1800 ng/mL
Assay Sensitivity 7.5 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 11093
Gene Symbol ADAMTS13
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target ADAMTS13
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.