Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADAMTS18 Partial ORF (NP_955387.1, 942 a.a. - 1047 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89932591 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-591 25 μg
89-932-590 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-932-591 Supplier Abnova™ Supplier No. H00170692Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has a high sequence similarity to the protein encoded by gene ADAMTS16, another family member. It is thought to function as a tumor suppressor. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined. [provided by RefSeq]

Sequence: TCSKACAGGQQSRKIQCVQKKPFQKEEAVLHSLCPVSTPTQVQACNSHACPPQWSLGPWSQCSKTCGRGVRKRELLCKGSAAETLPESQCTSLPRPELQEGCVLGR

Specifications

Accession Number NP_955387.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 170692
Molecular Weight (g/mol) 37.4kDa
Name ADAMTS18 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen TCSKACAGGQQSRKIQCVQKKPFQKEEAVLHSLCPVSTPTQVQACNSHACPPQWSLGPWSQCSKTCGRGVRKRELLCKGSAAETLPESQCTSLPRPELQEGCVLGR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ADAMTS21
Common Name ADAMTS18
Gene Symbol ADAMTS18
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.