Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC Human Adenovirus 37, Strain GW (76-19026) - Frozen viral stock (ATCC VR-929)

Supplier:  ATCC VR929

Encompass

Human adenovirus 37 (strain GW (76-19026)) is a Mastadenovirus supplied as a frozen viral stock intended for infectious and respiratory disease research. The strain was isolated from a human case of keratoconjunctivitis and is propagated in WI-38 human diploid fibroblast cells.

  • Strain: GW (76-19026).
  • Propagation host: WI-38 human diploid fibroblast cells.
  • Isolation source: human eye with keratoconjunctivitis.
  • Geographic origin: Netherlands.
  • Product format: frozen viral stock.
  • Storage: -70°C or colder.
  • Biosafety level: BSL-2.
  • Applications: infectious and respiratory disease research.

Catalog No. NC3834022


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.