Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADIPOR1 Partial ORF (NP_057083.2, 72 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89932718 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-718 25 μg
89-932-717 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-932-718 Supplier Abnova™ Supplier No. H00051094Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The adiponectin receptors, ADIPOR1 and ADIPOR2 (MIM 607946), serve as receptors for globular and full-length adiponectin (MIM 605441) and mediate increased AMPK (see MIM 602739) and PPAR-alpha (PPARA; MIM 170998) ligand activities, as well as fatty acid oxidation and glucose uptake by adiponectin (Yamauchi et al., 2003 [PubMed 12802337]).[supplied by OMIM]

Sequence: QAHHAMEKMEEFVYKVWEGRWRVIPYDVLPDWLKDNDYLLHGHRPPMPSFRACFKSIFRIHTETG

Specifications

Accession Number NP_057083.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51094
Molecular Weight (g/mol) 32.89kDa
Name ADIPOR1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen QAHHAMEKMEEFVYKVWEGRWRVIPYDVLPDWLKDNDYLLHGHRPPMPSFRACFKSIFRIHTETG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ACDCR1/CGI-45/CGI45/FLJ25385/FLJ42464/PAQR1/TESBP1A
Common Name ADIPOR1
Gene Symbol ADIPOR1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.