Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human AIF-1/Iba1 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP266674 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266674 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266674 Supplier Novus Biologicals™ Supplier No. NBP266674

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human AIF-1/Iba1 ELISA Kit (Chemiluminescence) measures human AIF-1/Iba1 in serum, plasma and other biological fluids.

Specifications

Assay Range 12.50 - 800 pg/mL
Assay Sensitivity 7.50 pg/mL
Cross Reactivity This kit recognizes Human AIF1 in samples. No significant cross-reactivity or interference between Human AIF1 and analogues was observed.
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
Specificity No significant cross-reactivity or interference between Human AIF1 and analogues was observed for this Human AIF-1/Iba1 ELISA Kit (Chemiluminescence).
For Use With (Application) ELISA
Gene ID (Entrez) 199
Gene Symbol AIF1
Host Species Human
Quantity 1 Kit
Research Discipline Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1
Target AIF-1/Iba1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.