Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Airway Trypsin-like Protease/HAT Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
SDP

Catalog No. RD10000916
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000916 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10000916 Supplier R&D Systems Supplier No. FAB2695G100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Airway Trypsin-like Protease/HAT/TMPRSS11D Monoclonal antibody specifically detects Airway Trypsin-like Protease/HAT/TMPRSS11D in Human samples. It is validated for Western Blot

Specifications

Antigen Airway Trypsin-like Protease/HAT/TMPRSS11D
Applications Western Blot
Classification Monoclonal
Clone 337029
Conjugate Alexa Fluor 488
Formulation Supplied 0.2 mg/mL in 1X PBS with RDF1 and 0.09% Sodium Azide
Gene Alias airway trypsin like protease, Airway trypsin-like protease, EC 3.4.21, EC 3.4.21.-, HAT, MGC150587, MGC150588, TMPRSS11D, transmembrane protease serine 11D, transmembrane protease, serine 11D
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Airway Trypsin-like Protease/HAT, Ala42-Ile418, Accession # O60235
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9407
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8 °C as supplied
Product Type Antibody
Form Purified
Isotype IgG1
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.