Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human AITRL Recombinant Protein, PeproTech®

Catalog No. 3102220UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
310-22-20UG 20 μg
31022100UG 100 μg
310-22-1MG 1 mg
31022250UG 250 μg
31022500UG 500 μg
310-22-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 310-22-20UG Supplier Gibco™ Supplier No. 3102220UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human soluble AITRL is a 14.4 kDa protein, containing 127 amino acid residues corresponding to the extracellular domain of AITRL. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The tumor necrosis factor (TNF) and TNF receptor (TNFR) gene superfamilies regulate numerous biological functions including cell proliferation, differentiation, and survival through regulating the activation of the transcription factor NF-kappa-B and various mitogen-activated protein kinases. The glucocorticoid-induced tumor necrosis factor receptor (GITR) is an emerging member of this family that is expressed on CD4+ CD25+ regulatory T cells and appears to have crucial immune regulation functions. Its ligand GITRL is expressed in endothelial and antigen-presenting cells and can activate NF-kappa-B, induce both pro- and anti-apoptotic effects, inhibit the suppressive activity of regulatory T cells, and co-stimulate responder T cells through GITR. Dominant negative forms of NIK and TRAF2 expressed in transfected 293 cells substantially inhibited NF-kappa-B activation, suggesting that the GITRL-GITR pathway involves both NIK and TRAF2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UNG2
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 8995
Molecular Weight (g/mol) 14.3 kDa
Name Human AITRL
Quantity 20 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Activation-inducible TNF-related ligand; AITR ligand; AITRL; GITR ligand; Gitrl; GITRLigand; Glucocorticoid-induced TNF-related ligand; glucocorticoid-induced TNFR-related protein ligand; glucocorticoid-induced-tumor necrosis factor receptor ligand; hGITRL; RP1-15D23.1; TL6; TNF superfamily member 18; Tnfsf18; TNLG2A; tumor necrosis factor (ligand) superfamily member 18; tumor necrosis factor (ligand) superfamily, member 18; tumor necrosis factor ligand 2 A; tumor necrosis factor ligand superfamily member 18; tumor necrosis factor superfamily member 18; UNQ149/PRO175
Common Name GITRL
Gene Symbol TNFSF18
Biological Activity Determined by its ability to stimulate IL-8 production by human PBMC using a concentration range of 1.0-10.0 ng/ml Please Note: Results may vary with PBMC donors.
Conjugate Unconjugated
Recombinant Recombinant
Sequence METAKEPCMA KFGPLPSKWQ MASSEPPCVN KVSDWKLEIL QNGLYLIYGQ VAPNANYNDV APFEVRLYKN KDMIQTLTNK SKIQNVGGTY ELHVGDTIDL IFNSEHQVLK NNTYWGIILL ANPQFIS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.