Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AKAP4 Partial ORF (NP_003877, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89932982 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-982 25 μg
89-932-981 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-932-982 Supplier Abnova™ Supplier No. H00008852Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is localized to the sperm flagellum and may be involved in the regulation of sperm motility. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MMAYSDTTMMSDDIDWLRSHRGVCKVDLYNPEGQQDQDRKVICFVDVSTLNVEDKDYKDAASSSSEGNLNLGSLEEKEIIVIKDTEKKDQSKTEGSVCLF

Specifications

Accession Number NP_003877
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8852
Molecular Weight (g/mol) 36.74kDa
Name AKAP4 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MMAYSDTTMMSDDIDWLRSHRGVCKVDLYNPEGQQDQDRKVICFVDVSTLNVEDKDYKDAASSSSEGNLNLGSLEEKEIIVIKDTEKKDQSKTEGSVCLF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AKAP82/FSC1/HI/hAKAP82/p82
Common Name AKAP4
Gene Symbol AKAP4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.