Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AKR1C3 (NP_003730, 1 a.a. - 323 a.a.) Full-length Recombinant Protein with His tag

Catalog No. 89933022 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-022 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-022 Supplier Abnova™ Supplier No. P3481
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols by utilizing NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme catalyzes the reduction of prostaglandin (PG) D2, PGH2 and phenanthrenequinone (PQ), and the oxidation of 9alpha,11beta-PGF2 to PGD2. It may play an important role in the pathogenesis of allergic diseases such as asthma, and may also have a role in controlling cell growth and/or differentiation. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14. [provided by RefSeq]

Sequence: MGSSHHHHHHSSGLVPRGSHMDSKHQCVKLNDGHFMPVLGFGTYAPPEVPRSKALEVTKLAIEAGFRHIDSAHLYNNEEQVGLAIRSKIADGSVKREDIFYTSKLWSTFHRPELVRPALENSLKKAQLDYVDLYLIHSPMSLKPGEELSPTDENGKVIFDIVDLCTTWEAMEKCKDAGLAKSIGVSNFNRRQLEMILNKPGLKYKPVCNQVECHPYFNRSKLLDFCKSKDIVLVAYSALGSQRDKRWVDPNSPVLLEDPVLCALAKKHKRTPALIALRYQLQRGVVVLAKSYNEQRIRQNVQVFEFQLTAEDMKAIDGLDRNLHYFNSDSFASHPNYPYSDEY

Specifications

Accession Number NP_003730
Concentration 1 mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation In 20mM Tris-HCl, pH 8.0 (10% glycerol)
Gene ID (Entrez) 8644
Molecular Weight (g/mol) 39kDa
Name AKR1C3 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing Loading 3 ug protein in 15% SDS-PAGE
Quantity 100 μg
Storage Requirements Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg of protein (determined by LAL method)
Gene Alias DD3/DDX/HA1753/HAKRB/HAKRe/HSD17B5/KIAA0119/hluPGFS
Common Name AKR1C3
Gene Symbol AKR1C3
Biological Activity Specific activity: approximately > 0.1 units/mg. Enzymatic activity was confirmed by measuring the amount of enzyme catalyzing the oxidation of 1μmole NADPH per minute at 25°C. Specific activity was expressed as units/mg protein.
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.