Learn More
Sino Biological Human AKT1 MAb 100UL 110547-R0002-100
Supplier:  Sino Biological 110547R0002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the residues around Ser308 of Human Phospho-Akt.
Catalog No. 50-241-2497
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.