Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AKT2 (NM_001626) Full-length Recombinant Protein with His tag expressed in Sf9 cells

Catalog No. 89933056 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-056 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-056 Supplier Abnova™ Supplier No. P3837

Used for Func, SDS-PAGE

This gene is a putative oncogene encoding a protein belonging to a subfamily of serine/threonine kinases containing SH2-like (Src homology 2-like) domains. The gene was shown to be amplified and overexpressed in 2 of 8 ovarian carcinoma cell lines and 2 of 15 primary ovarian tumors. Overexpression contributes to the malignant phenotype of a subset of human ductal pancreatic cancers. The encoded protein is a general protein kinase capable of phophorylating several known proteins. [provided by RefSeq]

Specifications

Accession Number NM_001626
For Use With (Application) Functional Study, SDS-PAGE
Formulation In 50mM Tris-HCl, 150mM NaCl, pH 7.5 (0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol)
Gene ID (Entrez) 208
Molecular Weight (g/mol) 58kDa
Name AKT2 (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PKBB/PKBBETA/PRKBB/RAC-BETA
Common Name AKT2
Gene Symbol AKT2
Biological Activity The specific activity was determined to be 46nmol/min/mg as per Activity Assay Protocol.
Species Baculovirus
Recombinant Recombinant
Protein Tag His
Expression System Insect cell (Sf9) expression system
Form Liquid
Purity or Quality Grade >75% by densitometry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.