Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human albumin Goat anti-Human, HRP, Polyclonal, Nordic
SDP

Catalog No. 502100800
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-0800 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-0800 Supplier Vector Laboratories Supplier No. GAHU/ALB/PO
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the antiserum. Diluted antiserum should be stored at +4 C, not refrozen, and preferably used the same day.

Human albumin Polyclonal antibody specifically detects Human albumin in Human samples. It is validated for Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Human albumin
Applications Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 5-10 mg/mL
Conjugate HRP
Formulation PBS with no preservative; pH
Gene ALB
Gene Accession No. P02768
Gene Alias ALB; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341
Host Species Goat
Immunogen Human albumin
Purification Method Protein G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.