Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ALK-6/BMPR-IB ELISA Kit

Catalog No. EH18RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH18RBX10 10 x 96 Tests
EH18RBX5 5 x 96 Tests
EH18RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH18RBX10 Supplier Invitrogen™ Supplier No. EH18RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human ALK-6/BMPR-IB quantitates human ALK-6/BMPR-IB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ALK-6/BMPR-IB.

BMPR1B (also known as ALK6, CDw293) is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor, which is closely related to the activin receptors, ACVR1 and ACVR2. BMPR1B is mainly involved in the endochondral bone formation and embryogenesis. BMPR1B expressed in normal and cancerous prostate tissues and used in the endocrine therapy that given to the prostate cancer patients. BMPR1B receptor trafficking also play a significant role in FOP pathogenesis and used in human T-cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number activin receptor-like kinase 6; Acvrlk6; AI385617; ALK6; ALK-6; AMDD; AV355320; BDA1D; BDA2; BMP type-1 B receptor; BMP15; Bmpr1b; BMPR-1 B; BMPRIB; BMPR-IB; bone morphogenetic protein binding serine/threonine kinase receptor; bone morphogenetic protein receptor IB; bone morphogenetic protein receptor type 1 B; bone morphogenetic protein receptor type IB; bone morphogenetic protein receptor type-1 B; bone morphogenetic protein receptor, type 1 B; bone morphogenetic protein receptor, type IB; CDw293; CFK-43 A; serine/threonine receptor kinase; serine/threonine-protein kinase receptor R6; SKR6
Assay Range 24 to 6,000 pg/mL
Assay Sensitivity 24 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 658
Gene Symbol BMPR1B
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CDw293
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.