Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ALK (BAG10812.1, 1058 a.a. - 1620 a.a.) C1156Y mutant Partial Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells

Catalog No. 89933173 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-173 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-173 Supplier Abnova™ Supplier No. P5736
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

The 2;5 chromosomal translocation is frequently associated with anaplastic large cell lymphomas (ALCLs). The translocation creates a fusion gene consisting of the ALK (anaplastic lymphoma kinase) gene and the nucleophosmin (NPM) gene: the 3' half of ALK, derived from chromosome 2, is fused to the 5' portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK (leukocyte tyrosine kinase). ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system. [provided by RefSeq]

Specifications

Accession Number BAG10812.1
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 238
Molecular Weight (g/mol) 90kDa
Name ALK (C1156Y) (Human) Recombinant Protein
Preparation Method Baculovirus infected insect cell (Sf21) expression system
Purification Method Glutathione sepharose chromatography
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CD246/Ki-1/TFG/ALK
Common Name ALK
Gene Symbol ALK
Biological Activity The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : Srctide. ATP: 100 μM.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Baculovirus infected insect cell (Sf21) expression system
Form Liquid
Purity or Quality Grade 71% by SDS-PAGE/CBB staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.