Learn More
Human Alkaline Phosphatase/ALPL Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Alkaline Phosphatase/ALPL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF |
| Classification | Monoclonal |
| Clone | B4-78 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Alias | Akp2, Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALP, TNSALPMGC167935 |
| Gene Symbols | ALPL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.