Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human alpha-Synuclein ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP262771 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262771 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262771 Supplier Novus Biologicals™ Supplier No. NBP262771
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human alpha-Synuclein ELISA Kit (Chemiluminescence) measures human alpha-Synuclein in serum, plasma and other biological fluids.

Specifications

Assay Range 12.50-800 pg/mL
Assay Sensitivity 7.50 pg/mL
Cross Reactivity This kit recognizes Human SNC(alpha) in samples. No significant cross-reactivity or interference between Human SNC(alpha) and analogues was observed.
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
Specificity This Human alpha-Synuclein ELISA Kit (Chemiluminescence) is specific for Human alpha-Synuclein in samples. No significant cross-reactivity or interference between Human alpha-Synuclein and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 6622
Gene Symbol SNCA
Host Species Human
Quantity 1 Kit
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target alpha-Synuclein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.