Learn More
Bon Opus Biosciences Recombinant Alkaline Phosphatase - Source: Human 293 Cells
SDP
Supplier:  Bon Opus Biosciences BP23950UG
Encompass_Preferred
Recombinant Human Alkaline Phosphatase is produced by our Mammalian expression system and the target gene encoding Ile23-Asp506 is expressed with a 6His tag at the C-terminus. Accession#P05187. Formulation: Supplied as a 0.2 um filtered solution of 20mM Tris-Hcl, 150mM NaCl, 0.2mM CaCl2, 10% Glycerol, pH7.5
Catalog No. NC2733672
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.