Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AMBN Full-length ORF (NP_057603.1, 1 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89933281 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-281 10 μg
89-933-282 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-933-281 Supplier Abnova™ Supplier No. H00000258P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Ameloblastin is thought to represent an unique ameloblast-specific gene product that may be important in enamel matrix formation and mineralization. The gene is located on chromosome 4 near other genes associated with mineralized tissues: osteopontin, bone sialoprotein, and bone morphogenetic protein 3. Based on its cytogenetic location, this gene is a candidate gene for one form of the disorder, dentinogenesis imperfecta, and/or the disorder, autosomal dominant amylogenesis imperfecta. [provided by RefSeq]

Sequence: MSASKIPLFKMKDLILILCLLEMSFAVPFFPQQSGTPGMASLSLETMRQLGSLQRLNTLSQYSRYGFGKSFNSLWMHGLLPPHSSLPWMRPREHETQQYEYSLPVHPPPLPSQPSLKPQQPGLKPFLQSAAATTNQATALKEALQPPIHLGHLPLQEGELPLVQQQVAPSDKPPKPELPGVDFADPQGPSLPGMDFPDPQGPSLPGLDFADPQGSTIFQIARLISHGPMPQNKQSPLYPGMLYVPFGANQLNAPARLGIMSSEEVAGGREDPMAYGAMFPGFGGMRPGFEGMPHNPAMGGDFTLEFDSPVAATKGPENEEGGAQGSPMPEANPDNLENPAFLTELEPAPHAGLLALPKDDIPGLPRSPSGKMKGLPSVTPAAADPLMTPELADVYRTYDADMTTSVDFQEEATMDTTMAPNSLQTSMPGNKAQEPEMMHDAWHFQEP

Specifications

Accession Number NP_057603.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 258
Molecular Weight (g/mol) 74.7kDa
Name AMBN (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name AMBN
Gene Symbol AMBN
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.