Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AmiGO2 ELISA Kit

Catalog No. EH23RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH23RB 96 Tests
EH23RBX5 5 x 96 Tests
EH23RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH23RB Supplier Invitrogen™ Supplier No. EH23RB
Add to Cart
Add to Cart

ELISA

Human AmiGO2 quantitates human AmiGO2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human AmiGO2.

A novel primary response gene, Alivin 1 (ali1), is an activity-dependent gene and promotes survival of neurons. Sequence analyses reveal that rat, mouse, and human Ali1 proteins contain seven leucine-rich repeats, one IgC2-like loop and a transmembrane domain, and display homology to Kek and Trk families. Results suggest that expression of ali1 promotes depolarization-dependent survival of the granule neuron. Mouse ali1 was mapped to a locus approximately 55.3 cM from the centromere on chromosome 15 that is syntenic to positional candidate loci for familial Alzheimer's disease type 5 and Parkinson's disease 8 on human chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number adhesion molecule with Ig like domain 2; adhesion molecule with Ig-like domain 2; AI415330; ALI1; alivin 1; alivin-1; AMIGO2; AMIGO-2; amigo2 {ECO:0000312; amphoterin induced gene 2; amphoterin induced gene and ORF 2; amphoterin-induced protein 2; AW208913; DEGA; differentially expressed in gastric adenocarcinoma; differentially expressed in gastric adenocarcinomas; EMBL:AAO48951.1}; transmembrane protein AMIGO2
Assay Range 0.2 to 50 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 347902
Gene Symbol Amigo2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Alivin 1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.