Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human AMPKβ2 Antibody, Novus Biologicals™

Catalog No. MAB11639100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11639100 100 μg
MAB11639SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB11639100 Supplier R&D Systems Supplier No. MAB11639100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

AMPK beta 2/PRKAB2 Monoclonal antibody specifically detects AMPK beta 2/PRKAB2 in Human samples. It is validated for Western Blot,Simple Western

Specifications

Antigen AMPK beta 2/PRKAB2
Applications Western Blot, Simple Western
Classification Monoclonal
Clone 1092615
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Simple Western 20 μg/mL
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. O43741
Gene Alias 5'-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5'-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, PRKAB2, protein kinase, AMP-activated, beta 2 non-catalytic subunit
Host Species Mouse
Immunogen E. coli - derived recombinant human AMPKbeta2, Met1-Ile272, Accession # O43741
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5565
Reconstitution Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°CC as supplied. 1 month, 2 to 8°CC under sterile conditions after reconstitution. 6 months, -20 to -70°CC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.