Learn More
TRIBIOSCIENCE INC Human Amyloid Beta 40 / Aβ40 ELISA Kit, 1 x 96-well plate
SDP
Supplier:  TRIBIOSCIENCE INC TBS3298
Encompass_Preferred
The Human Aβ40 ELISA is designed to quantitatively detect human Aβ40 levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 31 to 2000 pg/mL.
Catalog No. NC2886062
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.