Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Amyloid beta (aa1-40) Quantikine ELISA Kit
SDP

Catalog No. DAB140B Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DAB140B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DAB140B Supplier R&D Systems™ Supplier No. DAB140B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Amyloid beta (aa1-40) Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Amyloid beta (aa1-40) Quantikine ELISA Kit quantifies human beta Amyloid in cell culture supernates (200 ul), tissue lysates (10 ul), cerebrospinal fluid (10 ul).

Specifications

Assay Range 15.60 to 1000 pg/mL
Assay Sensitivity 8.17 pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Cerebrospinal Fluid, Tissue Lysates
Specificity Natural Human Amyloid beta (aa1-40)
For Use With (Application) ELISA
Gene ID (Entrez) 351
Gene Symbol APP
Quantity 1 Kit
Sample Volume Cell Culture Supernates (200uL),Tissue Lysates (10uL),Cerebrospinal Fluid (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym Abeta, Amyloid beta, beta-amyloid peptide
Target beta Amyloid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.