Learn More
IBL America Amyloid Beta (1-38) (FL) Aβ (Human) ELISA
Supplier:  IBL America 27717
Encompass_Preferred
Solid phase sandwich ELISA for the determination human amyloid beta (1-38) which held N terminal side completely in cerebrospinal fluids and cell culture supernatant. For research use only, not for use in diagnostic procedures.
Catalog No. 50-212-0347