Learn More
Abnova™ Human ANAPC1 Partial ORF (NP_073153, 1841 a.a. - 1944 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
ANAPC1 is 1 of at least 10 subunits of the anaphase-promoting complex (APC), which functions at the metaphase-to-anaphase transition of the cell cycle and is regulated by spindle checkpoint proteins. The APC is an E3 ubiquitin ligase that targets cell cycle regulatory proteins for degradation by the proteasome, thereby allowing progression through the cell cycle.[supplied by OMIM]
Specifications
Specifications
| Accession Number | NP_073153 |
| For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 64682 |
| Molecular Weight (g/mol) | 37.18kDa |
| Name | ANAPC1 (Human) Recombinant Protein (Q01) |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | NSEFLPVVKCTIDNTLDQWLQVGGDMCVHAYLSGQPLEESQLSMLACFLVYHSVPAPQHLPPIGLEGSTSFAELLFKFKQLKMPVRALLRLAPLLLGNPQPMVM |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.