Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Human Androgen Receptor Peptide, Synthetic, 90% Purity, 1 mg

Catalog No. 502704279
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4279 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4279 Supplier Abcam Supplier No. AB2230991MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide for use in blocking applications, with over 90% purity.

  • Form: Lyophilized
  • Purity: >90%
  • Tested Applications: BL
  • Storage Conditions: Store at -20°C
  • Shipping Temperature: Blue Ice
  • Formulation: 30% Trifluoroacetic acid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Androgen Receptor (AR-V7 Specific) , Human
Substrate Properties Peptide-based Substrate
Shipping Condition Blue Ice
Reactivity Human
Label or Dye Unconjugated
Buffer 30% trifluoroacetic acid
Content And Storage Store at -20°C.
Detection Location In-Blot Detection
Form Lyophilized
Cross Reactivity Human
Formulation 30% Trifluoroacetic acid
Quantity 1 mg
For Use With (Application) Blocking
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.